2-Methoxy-4-nitrobenzonitril 98%

Lieferant: Thermo Scientific
Achtung

440220050
ACRO440220050EA 104 EUR
ACRO440220050
2-Methoxy-4-nitrobenzonitril 98%
2-Methoxy-4-nitrobenzonitril
Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093781
CAS-Nummer: 101084-96-2
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR