O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, (Benzotriazol-1-yloxy)dipyrrolidinocarbenium hexafluorophosphate, (1H-Benzo[d][1,2,3]triazol-1-yl)(di(pyrrolidin-1-yl)methylene)oxonium hexafluorophosphate(V)

440250250
ACRO440250250EA 496 EUR
ACRO440250250
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate 98%
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Formel: C₁₅H₂₀F₆N₅OP
Molecular Weight: 431,323 g/mol
Schmelzpunkt: 176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191770
CAS-Nummer: 105379-24-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 15.83 to 16.64 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR