N,N,N',N'-Tetramethyl-S- (1-oxido-2-pyridyl) thiuroniumhexafluorophosphat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate

440260050
ACRO440260050EA 78.2 EUR
ACRO440260050
N,N,N',N'-Tetramethyl-S- (1-oxido-2-pyridyl) thiuroniumhexafluorophosphat 97%
N,N,N',N'-Tetramethyl-S- (1-oxido-2-pyridyl) thiuroniumhexafluorophosphat
Formel: C₁₀H₁₆F₆N₃OPS
Molecular Weight: 371,28 g/mol
Schmelzpunkt: 111,7…113.6 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD05865266
CAS-Nummer: 212333-72-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Elemental Analysis 10.64 to 12.00 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR