Di-tert-butyl hydrazodicarboxylate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
440420100
ACRO440420100EA
98.8
EUR
ACRO440420100
Di-tert-butyl hydrazodicarboxylate 97%
Di-tert-butyl hydrazodicarboxylate
Formel:
C₁₀H₂₀N₂O₄ Schmelzpunkt: 120…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015000 CAS-Nummer: 16466-61-8 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...