1-(Boc-amino)cyclobutanecarboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-BOC-1-Aminocyclobutanecarboxylic acid, Boc-cyclovaline, N-tert-Butoxycarbonyl-1-aminocyclobutanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, N-Boc-1-aminocyclobutanecarboxylic acid
440440050
ACRO440440050EA
256
EUR
ACRO440440050
1-(Boc-amino)cyclobutanecarboxylic acid 97%
1-(Boc-amino)cyclobutanecarboxylic acid
Formel:
C₁₀H₁₇NO₄ Schmelzpunkt: 129…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682623 CAS-Nummer: 120728-10-1 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...