N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 97%
Lieferant: Thermo Scientific
Synonyme:
(S)-tert-Butyl (1-(methoxy(methyl)amino)-3-methyl-1-oxobutan-2-yl)carbamate, (S)-[1-(Methoxy-methyl-carbamoyl)-2-methyl-propyl]-carbamic acid-tert-butyl ester, N2-{[(1,1-dimethylethyl)oxy]carbonyl}-N1-methyl-N1-(methyloxy)-L-valinamide, tert-butyl (S)-(1-(methoxy(methyl)amino)-3-methyl-1-oxobutan-2-yl)carbamate
440580050
ACRO440580050EA
161
EUR
ACRO440580050
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 97%
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide
Formel:
C₁₂H₂₄N₂O₄ Molecular Weight: 260,334 g/mol Siedepunkt: 248 °C (760 mmHg) Schmelzpunkt: 34…37 °C Flammpunkt: 113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151897 CAS-Nummer: 87694-52-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Low melting solid or liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | -15° to -20° (20°C, 589 nm) (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...