Xylencyanol FF ∼75% ~75%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Acid Blue 147, Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium
440700100
440700500
ACRO440700100EA
48.8
EUR
ACRO440700100
ACRO440700500
Xylencyanol FF ∼75% ~75%
Xylencyanol FF
Formel:
C₂₅H₂₇N₂NaO₆S₂ Molecular Weight: 538.62 g/mol Schmelzpunkt: 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040657 CAS-Nummer: 2650-17-1 |
Spezifikation Testergebnisse
Appearance | Blue-grey to green Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % (105°C) |
Solubility | (0.1% in water) Dark blue |
Dye content | ≥70 % |
UV Lambda max | 612 to 616 nm |
Suitability | (as track. dye for DNA seq) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...