Xylencyanol FF ∼75% ~75%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Acid Blue 147, Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium

440700100 440700500
ACRO440700100EA 48.8 EUR
ACRO440700100 ACRO440700500
Xylencyanol FF ∼75% ~75%
Xylencyanol FF
Formel: C₂₅H₂₇N₂NaO₆S₂
Molecular Weight: 538.62 g/mol
Schmelzpunkt: 295 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040657
CAS-Nummer: 2650-17-1

Bestellen

Spezifikation Testergebnisse

Appearance Blue-grey to green Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
Solubility (0.1% in water) Dark blue
Dye content ≥70 %
UV Lambda max 612 to 616 nm
Suitability (as track. dye for DNA seq) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR