Boc-L-valine hydroxysuccinimide ester 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Boc-L-valine N-succinimidyl ester, Boc-Val-OSu
440920050
ACRO440920050EA
56
EUR
ACRO440920050
Boc-L-valine hydroxysuccinimide ester 98%
Boc-L-valine hydroxysuccinimide ester
Formel:
C₁₄H₂₂N₂O₆ Molecular Weight: 314,338 g/mol Schmelzpunkt: 126…129 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00037906 CAS-Nummer: 3392-12-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % |
HPLC | ≥97.5 % |
Specific optical rotation | -36.0° to -40.0° (20°C, 589 nm) (c=1 in dioxane) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...