3-(Triphenylphosphoranylidene)tetrahydropyran-2-one 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Triphenylphosphoranylidene)tetrahydro-2H-pyran-2-one, 2H-Pyran-2-one, tetrahydro-3-(triphenylphosphoranylidene)-
440980010
ACRO440980010EA
177
EUR
ACRO440980010
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one 97%
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one
Formel:
C₂₃H₂₁O₂P Molecular Weight: 360,385 g/mol Schmelzpunkt: 228…230 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD20481847 CAS-Nummer: 218282-19-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...