N-Methylpyrrole-3-boronic acid pinacol ester 90%

Lieferant: Thermo Fisher Scientific
Achtung

441000010 441000050
ACRO441000010EA 167 EUR
ACRO441000010 ACRO441000050
N-Methylpyrrole-3-boronic acid pinacol ester 90%
N-Methylpyrrole-3-boronic acid pinacol ester
Formel: C₁₁H₁₈BNO₂
Schmelzpunkt: 44…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13182200
CAS-Nummer: 953040-54-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥88.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR