N-Methylpyrrole-2-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

441040010
ACRO441040010EA 66.5 EUR
ACRO441040010
N-Methylpyrrole-2-boronic acid pinacol ester 97%
N-Methylpyrrole-2-boronic acid pinacol ester
Formel: C₁₁H₁₈BNO₂
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659917
CAS-Nummer: 850567-47-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
Melting point 45 to 50 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR