(S)-N-Fmoc-α-methyl-2-fluorophenylalanine 98%, 98% ee
Lieferant: Thermo Scientific
Synonyme:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)-2-methylpropanoic acid, FMOC-α-ME-L-PHE(2-F)-OH, FMOC-α-METHYL-D-2-FLUOROPHENYLALANINE
441095000
ACRO441095000EA
533
EUR
ACRO441095000
(S)-N-Fmoc-α-methyl-2-fluorophenylalanine 98%, 98% ee
(S)-N-Fmoc-alpha-methyl-2-fluorophenylalanine
Formel:
C₂₅H₂₂FNO₄ Molecular Weight: 419,452 g/mol Schmelzpunkt: 122…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD17019320 CAS-Nummer: 1172127-44-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Enantiomeric excess | ≥97.5 % |
Specific optical rotation | -46° to -50° (20°C, 589 nm) (c=1 in DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...