2-Methyl-L-cysteine hydrochloride 98%, 98% ee

Lieferant: Thermo Fisher Scientific
Achtung

441101000
ACRO441101000EA 115 EUR
ACRO441101000
2-Methyl-L-cysteine hydrochloride 98%, 98% ee
2-Methyl-L-cysteine hydrochloride
Formel: C₄H₁₀ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08063384
CAS-Nummer: 148766-37-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Elemental Analysis 7.96 to 8.36 % (N)
Specific optical rotation +6° to +10° (20°C, 589 nm) (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR