(R)-N-Fmoc-α-methyl-2-fluorophenylalanine 98%, 98% ee

Lieferant: Thermo Scientific

Synonyme: FMOC-α-ME-L-PHE(2-F)-OH, FMOC-α-METHYL-D-2-FLUOROPHENYLALANINE

441115000
ACRO441115000EA 144 EUR
ACRO441115000
(R)-N-Fmoc-α-methyl-2-fluorophenylalanine 98%, 98% ee
(R)-N-Fmoc-alpha-methyl-2-fluorophenylalanine
Formel: C₂₅H₂₂FNO₄
Molecular Weight: 419,452 g/mol
Schmelzpunkt: 111…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17019319
CAS-Nummer: 193086-74-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +46° to +50° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR