(R)-N-Fmoc-α-methylvaline 98%, 98% ee

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2,3-dimethylbutanoic acid, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-methyl-L-isovaline, Fmoc-(R)-2-amino-2,3-dimethylbutanoicacid, Fmoc-D-α-Me-Val-OH

441145000
ACRO441145000EA 424 EUR
ACRO441145000
(R)-N-Fmoc-α-methylvaline 98%, 98% ee
(R)-N-Fmoc-alpha-methylvaline
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,418 g/mol
Schmelzpunkt: 37…53 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12031696
CAS-Nummer: 616867-28-8
UN: 3175
ADR: 4.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7.5° to -9.5° (20°C, 589 nm) (c=1 in CHCl3)
Residual solvents ≤10 % MTBE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR