(S)-N-Fmoc-α-methylvaline 98%, 98% ee

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-methyl-D-isovaline, Fmoc-α-Me-D-Val-OH, Fmoc-α-Me-Val-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2,3-dimethylbutanoic acid

441155000
ACRO441155000EA 424 EUR
ACRO441155000
(S)-N-Fmoc-α-methylvaline 98%, 98% ee
(S)-N-Fmoc-alpha-methylvaline
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353.418 g/mol
Schmelzpunkt: 37…53 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08752490
CAS-Nummer: 169566-81-8
UN: 3175
ADR: 4.1 II

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +7.5° to +9.5° (20°C, 589 nm) (c=1 in CHCl3)
Residual solvents ≤10.0 % MTBE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR