(S)-α-Methyltryptophan hemihydrate 98%, 98% ee
Lieferant: Thermo Fisher Scientific
Synonyme:
H-α-ME-L-TRP-OH 0.5H2O, (S)-α-METHYLTRYPTOPHAN 1/2H2O
441201000
ACRO441201000EA
180
EUR
ACRO441201000
(S)-α-Methyltryptophan hemihydrate 98%, 98% ee
(S)-alpha-Methyltryptophan hemihydrate
Formel:
C₁₂H₁₄N₂O₂·0.5H₂O Molecular Weight: 227.26 g/mol Schmelzpunkt: >231 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD11982865 CAS-Nummer: 16709-25-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Enantiomeric excess | ≥98.0 % |
Specific optical rotation | -14° to -18° (20°C, 589 nm) (c=0.9 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...