2-Methylleucine 98%, 98% ee
Lieferant: Thermo Scientific
Synonyme:
(S)-2-Amino-2,4-dimethylpentanoic acid, H-α-Me-Leu-OH, (S)-alpha-Methylleucine
441261000
ACRO441261000EA
144
EUR
ACRO441261000
2-Methylleucine 98%, 98% ee
2-Methylleucine
Formel:
C₇H₁₅NO₂ Molecular Weight: 145,202 g/mol Schmelzpunkt: >237 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01716279 CAS-Nummer: 105743-53-1 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Assay | 97.5 to 102.5 % |
Enantiomeric excess | ≥98.0 % |
Specific optical rotation | +33° to +39° (20°C, 589 nm) (c=1 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...