(S)-α-Allylglycine ethyl ester-p-toluenesulphonate 98%, 98% ee
Lieferant: Thermo Scientific
Synonyme:
(S)-Allylglycine ethyl ester p-toluenesulphonate, (S)-α-Allylglycine ethyl ester p-toluenesulphonate
441280010
ACRO441280010EA
236
EUR
ACRO441280010
(S)-α-Allylglycine ethyl ester-p-toluenesulphonate 98%, 98% ee
(S)-alpha-Allylglycine ethyl ester-p-toluenesulphonate
Formel:
C₁₄H₂₁NO₅S Molecular Weight: 315,384 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031686 CAS-Nummer: 1231709-21-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Enantiomeric excess | ≥98.0 % |
Specific optical rotation | +10° to +16° (20°C, 589 nm) (c=1 in CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...