6-(Hydroxymethyl)benzothiazole 97%
Lieferant: Thermo Scientific
Synonyme:
(1,3-Benzothiazol-6-yl)methanol, benzo[d]thiazol-6-ylmethanol, 6-Benzothiazolemethanol
441330010
ACRO441330010EA
101
EUR
ACRO441330010
6-(Hydroxymethyl)benzothiazole 97%
6-(Hydroxymethyl)benzothiazole
Formel:
C₈H₇NOS Molecular Weight: 165,21 g/mol Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972625 CAS-Nummer: 19989-66-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 103 to 106 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...