6-(Chloromethyl)benzothiazole 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-(Chloromethyl)-1,3-benzothiazole, Benzothiazole, 6-(chloromethyl)-, 6-(Chloromethyl)benzo[d]thiazole
441350010
ACRO441350010EA
193
EUR
ACRO441350010
6-(Chloromethyl)benzothiazole 97%
6-(Chloromethyl)benzothiazole
Formel:
C₈H₆ClNS Molecular Weight: 183,653 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12756627 CAS-Nummer: 149440-49-3 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...