PyOxim 97%

Lieferant: Thermo Fisher Scientific

Synonyme: (E)-(((1-Cyano-2-ethoxy-2-oxoethylidene)amino)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

441410050
ACRO441410050EA 93 EUR
ACRO441410050
PyOxim 97%
PyOxim
Formel: C₁₇H₂₉F₆N₅O₃P₂
Molecular Weight: 527.389 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18643381
CAS-Nummer: 153433-21-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR