2-Amino-4-chlorpyridin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
441460050
441460250
ACRO441460050EA
55.7
EUR
ACRO441460050
ACRO441460250
2-Amino-4-chlorpyridin 97%
2-Amino-4-chlorpyridin
Formel:
C₅H₅ClN₂ Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04113820 CAS-Nummer: 19798-80-2 |
Spezifikation Testergebnisse
Appearance | White to light brown Solid |
Infrared spectrum | Conforms |
Melting point | 127 to 134 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...