2-Amino-3-iodpyridin 97%

Lieferant: Thermo Scientific
Achtung

441480050
ACRO441480050EA 286 EUR
ACRO441480050
2-Amino-3-iodpyridin 97%
2-Amino-3-iodpyridin
Formel: C₅H₅IN₂
Molecular Weight: 220,01 g/mol
Schmelzpunkt: 89,6…90.6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01651885
CAS-Nummer: 104830-06-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder or needles
Infrared spectrum Conforms
Melting point 86 to 92 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR