Dextransulfat Natriumsalz, M.W. ∼ 6,500 - 10,000
Lieferant: Thermo Fisher Scientific
441490050
441490250
441491000
ACRO441490050EA
54.7
EUR
ACRO441490050
ACRO441490250
ACRO441491000
Dextransulfat Natriumsalz, M.W. ∼ 6,500 - 10,000
Dextran sulphate sodium salt
Formel:
(C₆H₇Na₃O₁₄S₃)n Lagertemperatur: Kühlschrank |
MDL:
MFCD00081551 CAS-Nummer: 9011-18-1 |
Spezifikation Testergebnisse
Appearance | White to light yellow to light brown Powder |
Loss on drying | ≤10 % |
Solubility | (10 % in water) Clear to slightly hazy colorless to light yellow |
Molecular weight | 6500 to 10000 |
Residue after ignition | 35 % to 50 % |
pH | 5 to 8 (1 % solution) |
Specific optical rotation | +75° to +105° (20°C, 589 nm) (6% in water) |
Specific viscosity | 0.018 to 0.032 (1.0M in NaCl, 25°C) |
Sulfur (S) | 16 to 20 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...