2-Phenylethanaminhydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phenethylamine Hydrochloride

441910250
ACRO441910250EA 70.1 EUR
ACRO441910250
2-Phenylethanaminhydrochlorid 98%
2-Phenylethylamine hydrochloride
Formel: C₈H₁₁N
Molecular Weight: 157,64 g/mol
Schmelzpunkt: 220…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012893
CAS-Nummer: 156-28-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR