Pyridine hydrobromide 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pyridinium Bromide
442132500
ACRO442132500EA
64.2
EUR
ACRO442132500
Pyridine hydrobromide 98%
Pyridine hydrobromide
Formel:
C₅H₅N Molecular Weight: 160.01 g/mol Schmelzpunkt: 217…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012801 CAS-Nummer: 18820-82-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Water | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...