2,6-Dichlor-4-(trifluormethoxy)anilin 96%
Lieferant: Thermo Scientific
|
Achtung
|
442300050
ACRO442300050EA
116
EUR
ACRO442300050
2,6-Dichlor-4-(trifluormethoxy)anilin 96%
2,6-Dichlor-4-(trifluormethoxy)anilin
Formel:
C₇H₄Cl₂F₃NO Molecular Weight: 246,02 g/mol Schmelzpunkt: 32…34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190128 CAS-Nummer: 99479-66-0 |
Spezifikation Testergebnisse
Appearance | Colorless to white to light brown Crystalline needles or low melting solid |
Infrared spectrum | Conforms |
Melting point | 30 to 35 °C |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...