6-Nitro-2,4-xylidin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,4-Dimethyl-6-nitroanilin
442440050
ACRO442440050EA
122
EUR
ACRO442440050
6-Nitro-2,4-xylidin 97%
6-Nitro-2,4-xylidin
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,18 g/mol Schmelzpunkt: 69,5…70.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007739 CAS-Nummer: 1635-84-3 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Light yellow to orange Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...