5-Chlor-2-fluorbenzoesäure 97%

Lieferant: Thermo Scientific
Achtung

442510050
ACRO442510050EA 99.8 EUR
ACRO442510050
5-Chlor-2-fluorbenzoesäure 97%
5-Chlor-2-fluorbenzoesäure
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665762
CAS-Nummer: 394-30-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 151 to 157 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR