D-Phenylalanin-methylester Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

442620050 442620250
ACRO442620050EA 43 EUR
ACRO442620050 ACRO442620250
D-Phenylalanin-methylester Hydrochlorid 98%
D-Phenylalanine methyl ester hydrochloride
Formel: C₁₀H₁₃NO₂
Schmelzpunkt: 157…159 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00066112
CAS-Nummer: 13033-84-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -32° to -38° (20°C, 589 nm) (c=1 in EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR