Boc-D-aspartic acid β-benzyl ester 98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-D-aspartic acid 4-benzyl ester, Boc-D-Asp(OBzl)-OH

442870050
ACRO442870050EA 131 EUR
ACRO442870050
Boc-D-aspartic acid β-benzyl ester 98%
Boc-D-aspartic acid beta-benzyl ester
Formel: C₁₆H₂₁NO₆
Schmelzpunkt: 101…105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00038255
CAS-Nummer: 51186-58-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +18° to +21° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR