2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 98% analytischer Standard

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium 2-mercaptoethanesulphonate

443150050 443150100 443150250
ACRO443150050EA 49.9 EUR
ACRO443150050 ACRO443150100 ACRO443150250
2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 98% analytischer Standard
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel: C₂H₅NaO₃S₂
Molecular Weight: 164.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007535
CAS-Nummer: 19767-45-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 to 102 % (On dried substance)
Loss on drying ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR