3-Nitroanthranilsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-nitrobenzoic acid
443180100
ACRO443180100EA
142
EUR
ACRO443180100
3-Nitroanthranilsäure 97%
3-Nitroanthranilsäure
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Schmelzpunkt: 204…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024261 CAS-Nummer: 606-18-8 |
Spezifikation Testergebnisse
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Melting point | 204 to 210 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...