5-Brom-2,4-dimethoxypyrimidin 97%
Lieferant: Thermo Scientific
|
Achtung
|
443390050
ACRO443390050EA
93.6
EUR
ACRO443390050
5-Brom-2,4-dimethoxypyrimidin 97%
5-Bromo-2,4-dimethoxypyrimidine
Formel:
C₆H₇BrN₂O₂ Molecular Weight: 219,0400 g/mol Siedepunkt: 125 °C (17Torr) Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038016 CAS-Nummer: 56686-16-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 62 to 67 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...