Clindamycin hydrochloride 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cleocin Hydrochloride, (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride, 7[S]-Chloro-7-deoxylincomycin, Clindamycin HCl
443510100
ACRO443510100EA
97.1
EUR
ACRO443510100
Clindamycin hydrochloride 97%
Clindamycin hydrochloride
Formel:
C₁₈H₃₃ClN₂O₅S·HCl Molecular Weight: 461,45 g/mol Schmelzpunkt: 156…159 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD07793327 CAS-Nummer: 21462-39-5 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤6 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...