3-(N-BOC-aminomethyl)-5-bromopyridine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Carbamic acid,N-[(5-bromo-3-pyridinyl)methyl]-, 1,1-dimethylethyl ester, 3-Bromo-5-{[(tert-butoxycarbonyl)amino]methyl}pyridine, tert-Butyl ((5-bromopyridin-3-yl)methyl)carbamate, tert-Butyl (5-bromopyridin-3-yl)methylcarbamate
444060010
444060050
ACRO444060010EA
212
EUR
ACRO444060010
ACRO444060050
3-(N-BOC-aminomethyl)-5-bromopyridine 97%
3-(N-Boc-aminomethyl)-5-bromopyridine
Formel:
C₁₁H₁₅BrN₂O₂ Schmelzpunkt: 84,5…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10700149 CAS-Nummer: 943722-24-5 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...