1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

444440010
ACRO444440010EA 174 EUR
ACRO444440010
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₁H₁₇BN₂O₃
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027064
CAS-Nummer: 1150561-76-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR