3-Carbamoylphenylboronic acid pinacol ester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
444450050
ACRO444450050EA
300
EUR
ACRO444450050
3-Carbamoylphenylboronic acid pinacol ester 97%
3-Carbamoylphenylboronic acid pinacol ester
Formel:
C₁₃H₁₈BNO₃ Schmelzpunkt: 182…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663886 CAS-Nummer: 188665-74-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...