4-(4-Chlorphenyl)anilin 97%

Lieferant: Thermo Fisher Scientific
Achtung

444460010 444460050
ACRO444460010EA 126 EUR
ACRO444460010 ACRO444460050
4-(4-Chlorphenyl)anilin 97%
4-(4-Chlorphenyl)anilin
Formel: C₁₂H₁₀ClN
Schmelzpunkt: 127…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00477989
CAS-Nummer: 135-68-2

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to orange to brown Solid
Infrared spectrum Conforms
Melting point 126 to 132 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR