4-(4-Chlorphenyl)anilin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
444460010
444460050
ACRO444460010EA
126
EUR
ACRO444460010
ACRO444460050
4-(4-Chlorphenyl)anilin 97%
4-(4-Chlorphenyl)anilin
Formel:
C₁₂H₁₀ClN Schmelzpunkt: 127…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00477989 CAS-Nummer: 135-68-2 |
Spezifikation Testergebnisse
Appearance | Off-white to orange to brown Solid |
Infrared spectrum | Conforms |
Melting point | 126 to 132 °C |
GC | ≥96 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...