2-Amino-5-chlorphenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Amino-5-chlorophenyl boronic acid pinacol ester, 4-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
444470010
444470050
ACRO444470010EA
233
EUR
ACRO444470010
ACRO444470050
2-Amino-5-chlorphenylboronsäurepinakolester 97%
2-Amino-5-chlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₇BClNO₂ Schmelzpunkt: 91…93 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06795672 CAS-Nummer: 1073371-77-3 |
Spezifikation Testergebnisse
Appearance | White to pink to tan Solid |
Infrared spectrum | Conforms |
Melting point | 89 to 95 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...