4-Amino-2-chlorphenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-2-chlorophenylboronic acid,pinacolester, 4-Amino-2-chlorophenylboronic acid, piancol ester

444480010 444480050
ACRO444480010EA 190 EUR
ACRO444480010 ACRO444480050
4-Amino-2-chlorphenylboronsäurepinakolester 97%
4-Amino-2-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Schmelzpunkt: 123…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09864184
CAS-Nummer: 877160-63-9

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR