4-Amino-2-fluorphenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

444490010 444490050
ACRO444490010EA 145 EUR
ACRO444490010 ACRO444490050
4-Amino-2-fluorphenylboronsäurepinakolester 97%
4-Amino-2-fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
Schmelzpunkt: 163…166 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09951877
CAS-Nummer: 819057-45-9

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 161 to 167 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR