4-Amino-3-fluorphenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester

444500010 444500050
ACRO444500010EA 209 EUR
ACRO444500010 ACRO444500050
4-Amino-3-fluorphenylboronsäurepinakolester 97%
4-Amino-3-fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
Schmelzpunkt: 101…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09033884
CAS-Nummer: 819058-34-9

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to tan Solid
Infrared spectrum Conforms
Melting point 100 to 105 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR