3-Amino-2-methylphenylboronsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
444540050
ACRO444540050EA
370
EUR
ACRO444540050
3-Amino-2-methylphenylboronsäurepinakolester 97%
3-Amino-2-methylphenylboronsäurepinakolester
Formel:
C₁₃H₂₀BNO₂ Molecular Weight: 233,12 g/mol Siedepunkt: 120…121 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11054038 CAS-Nummer: 882678-96-8 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Solid |
Infrared spectrum | Conforms |
Melting point | 117 to 122 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...