4-Amino-3-nitrophenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

444570050
ACRO444570050EA 154 EUR
ACRO444570050
4-Amino-3-nitrophenylboronsäurepinakolester 97%
4-Amino-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₄
Schmelzpunkt: 174…177 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06795680
CAS-Nummer: 833486-94-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Solid
Infrared spectrum Conforms
Melting point 174 to 180 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR