2-Aminopyridine-4-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

444580010
ACRO444580010EA 318 EUR
ACRO444580010
2-Aminopyridine-4-boronic acid pinacol ester 97%
2-Aminopyridine-4-boronic acid pinacol ester
Formel: C₁₁H₁₇BN₂O₂
Schmelzpunkt: 161…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607735
CAS-Nummer: 1195995-72-2

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Solid
Infrared spectrum Conforms
Melting point 158 to 164 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR