6-Brom-2-methyl-2H-indazol 97%

Lieferant: Thermo Scientific
Achtung

444610010
ACRO444610010EA 222 EUR
ACRO444610010
6-Brom-2-methyl-2H-indazol 97%
6-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,0600 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09870048
CAS-Nummer: 590417-95-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR